Learn More
Cayman Chemical 4oxo2Nonenal Fatty Acids and D
SDP
Catalog No. 502889088
Encompass_Preferred
A lipid peroxidation product; actively modifies histidine and lysine residues on proteins and causes protein cross-linking
Catalog No. 50-288-9088 Supplier Cayman Chemical Supplier No. 1018510 MG
A lipid peroxidation product; actively modifies histidine and lysine residues on proteins and causes protein cross-linking