Learn More
Cayman Chemical 4oxo2Nonenald3 GC MS Standards
SDP
Catalog No. 502889030
Encompass_Preferred
An internal standard for the quantification of 4-ONE by GC- or LC-MS
Catalog No. 50-288-9030 Supplier Cayman Chemical Supplier No. 10004174500 UG
An internal standard for the quantification of 4-ONE by GC- or LC-MS