Learn More
Cayman Chemical 4-oxo-2-Nonenal-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1313400-91-7, 1 mg
SDP
Catalog No. 502890734
Encompass_Preferred
An internal standard for the quantification of 4-ONE by GC- or LC-MS
Catalog No. 50-289-0734 Supplier Cayman Chemical Supplier No. 100041741 MG
An internal standard for the quantification of 4-ONE by GC- or LC-MS