Learn More
Cayman Chemical 4oxo2Nonenald3 GC MS Standards
SDP
Supplier:  Cayman Chemical 10004174100 UG
Encompass_Preferred
An internal standard for the quantification of 4-ONE by GC- or LC-MS
Catalog No. 50-288-7905