Learn More
C&G Containers 125mL Bottles L3 (Bottle with Cap Attached Only) | 160/case
SDP
Catalog No. NC1853544
Encompass_Preferred
These glass jars are used primarily for the collection of liquid media, for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics.
Catalog No. NC1853544 Supplier C&G Containers Supplier No. LFB004900000
These glass jars are used primarily for the collection of liquid media, for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics.