Learn More
Container & Packaging Supply 4 oz Natural-Colored HDPE Plastic Cylinder Round Bottle w/ 20-410 neck finish, 480/cs
SDP
Catalog No. NC1847850
Encompass_Preferred
Cylinder bottles have sharp square shoulders and a square base. High density polyethylene (HDPE) is marked with the resin identification code of 2. HDPE has a high tensile strength, good resistance to chemicals, good impact resistance, highly rigid, and highly scratch resistant. This item has a 20-410 neck finish.
Catalog No. NC1847850 Supplier Container & Packaging Supply Supplier No. B777NT
Cylinder bottles have sharp square shoulders and a square base. High density polyethylene (HDPE) is marked with the resin identification code of 2. HDPE has a high tensile strength, good resistance to chemicals, good impact resistance, highly rigid, and highly scratch resistant. This item has a 20-410 neck finish.