Learn More
MSC PIP Sterile Eyewash Solution, 4 oz, ANSI Z358.1-2009, 1 ea
Catalog No. NC2938967
Encompass
Sterile eyewash squeeze bottles are readily available for emergency flushing and decontamination. They are designed for quick access, making them easy to keep within a 10-second walk from hazardous materials. These personal disposable eyewash bottles are approved by ANSI Z358.1-2009, FDA 21 CFR 200.59 & 21 CFR 349.
  • Portable and readily available for use
  • Bottles contain a buffered, saline solution that is sterile and superior to tap water for emergency eye care
  • Blow-fill-seal bottles are tamper resistant, yet easily removed in an emergency
  • Ensured quality and lasting value with FDA-grade polyethylene bottles featuring lot numbers and expiration dates
  • Features a 36-month shelf life from date of manufacturing
Catalog No. NC2938967 Supplier MSC Supplier No. 40993347
Sterile eyewash squeeze bottles are readily available for emergency flushing and decontamination. They are designed for quick access, making them easy to keep within a 10-second walk from hazardous materials. These personal disposable eyewash bottles are approved by ANSI Z358.1-2009, FDA 21 CFR 200.59 & 21 CFR 349.
  • Portable and readily available for use
  • Bottles contain a buffered, saline solution that is sterile and superior to tap water for emergency eye care
  • Blow-fill-seal bottles are tamper resistant, yet easily removed in an emergency
  • Ensured quality and lasting value with FDA-grade polyethylene bottles featuring lot numbers and expiration dates
  • Features a 36-month shelf life from date of manufacturing