Learn More
US Plastic Corporation 4OZ STERILEWARE SCOOPS
SDP
Catalog No. NC0319136
Encompass
Sampling scoops are the perfect shape and size for taking samples, especially from wide-mouth containers. Designed to hold various capacities, the one-piece scoop is molded of white high impact polystyrene.
Catalog No. NC0319136 Supplier US Plastic Corporation Supplier No. 80130
Sampling scoops are the perfect shape and size for taking samples, especially from wide-mouth containers. Designed to hold various capacities, the one-piece scoop is molded of white high impact polystyrene.