Learn More
MSC Leviton 2521 277/480 VAC 20A NEMA L22-20P Industrial Twist Lock Plug 4 Poles, 5 Wires, 3 Phase, Self-Grounding, 0.595 to 0.895" Cord Diam, Black/White
Catalog No. 501298762
Encompass
Leviton 2521 277/480 VAC 20A NEMA L22-20P Industrial Twist Lock Plug 4 Poles, 5 Wires, 3 Phase, Self-Grounding, 0.595 to 0.895" Cord Diam, Black/White
Catalog No. 50-129-8762 Supplier MSC Supplier No. 74364746
Leviton 2521 277/480 VAC 20A NEMA L22-20P Industrial Twist Lock Plug 4 Poles, 5 Wires, 3 Phase, Self-Grounding, 0.595 to 0.895" Cord Diam, Black/White