Learn More
Hach Company Vial Adapter Set (4 pcs) for DR1900 Spectrophotometer
Supplier:  Hach Company 9609800
Encompass
This vial adapter set contains four interchangeable adapters used with the DR1900 spectrophotometer to ensure correct positioning of sample vials or cuvettes during measurements.
  • Includes four adapters labeled A, B, C, and D.
  • Provides adapters for different vial or cuvette sizes and orientations.
  • Designed for use with the DR1900 spectrophotometer.
  • Replaces worn or missing adapters to restore proper sample positioning.
Catalog No. NC2455882