Learn More
Myron L Company 4PII ULTRA CONDUCTIVITY METER
SDP
Catalog No. NC9312057
Encompass
Myron L; Ultrameter II Models 4P; TDS; Resistivity; 7.7L x 2.7W x 2.5H in.; 9V Alkaline Battery; OT: 0deg. to 55deg.C; Protection ratings IP67/NEMA 6; Cell material VALOX; 100 readings memory. Please note, this item is sold with a non-dated COA.
Catalog No. NC9312057 Supplier Myron L Company Supplier No. 4PII
Myron L; Ultrameter II Models 4P; TDS; Resistivity; 7.7L x 2.7W x 2.5H in.; 9V Alkaline Battery; OT: 0deg. to 55deg.C; Protection ratings IP67/NEMA 6; Cell material VALOX; 100 readings memory. Please note, this item is sold with a non-dated COA.