Learn More
Biocare Medical, LLC 4PLUS HRP1000 UNIVERSAL 100ML
SDP
Catalog No. 5082877
Encompass_Preferred
4plus HRP Universal Detection is a two-step detection designed for sensitivity and reliability. This detects either mouse or rabbit primary antibodies with a biotinylated secondary antibody followed by a Horseradish Peroxidase (HRP) label.
Catalog No. 50-828-77 Supplier Biocare Medical, LLC Supplier No. HP504US
4plus HRP Universal Detection is a two-step detection designed for sensitivity and reliability. This detects either mouse or rabbit primary antibodies with a biotinylated secondary antibody followed by a Horseradish Peroxidase (HRP) label.