Learn More
Biocare Medical, LLC 4PLUS SA-AP LABEL 25ML
SDP
Supplier:  Biocare Medical, LLC AP605H
Encompass_Preferred
4plus Streptavidin AP Label is the second step in a two-step detection designed for sensitivity and reliability. This detects a biotinylated secondary antibody with an Streptavidin-Alkaline Phosphatase (AP) conjugate.
Catalog No. 50-823-65