Learn More
Cayman Chemical 4-Quinolone-3-Carboxamide CB2 Ligand, Purity: ≥98%, CAS: 1314230-69-7, 10 mg
SDP
Catalog No. 502951599
Encompass_Preferred
A selective, high-affinity ligand of the CB2 receptor (Ki = 0.6 nM v. CB1 binding at a Ki >10,000 nM in vitro) that may behave as an inverse agonist; displays antinociceptive activity in a formalin test in mice at a dose of 6 mg/kg
Catalog No. 50-295-1599 Supplier Cayman Chemical Supplier No. 1109310 MG
A selective, high-affinity ligand of the CB2 receptor (Ki = 0.6 nM v. CB1 binding at a Ki >10,000 nM in vitro) that may behave as an inverse agonist; displays antinociceptive activity in a formalin test in mice at a dose of 6 mg/kg