Learn More
Medchemexpress LLC (4R)-4-[(1S)-1-fluoroethyl]-3-[2-[[(1S)-1-[4-methyl-5-[2-(trifluoromethyl)-4-pyridinyl]... | 1628805-46-8 | MFCD31630851 | 99.2% | 490.45 g·mol⁻¹ | C23H22F4N6O2 | 1 ML
SDP
Catalog No. 5000242503
Encompass_Preferred
IDH-305 is an orally available, mutant-selective, brain-penetrant inhibitor of isocitrate dehydrogenase 1 (IDH1) that targets R132 mutations. It is provided for research use in biochemical and cellular assays and is supplied as a 10 mM solution in DMSO (1 mL) or as solid quantities.
  • Potent, mutant-selective inhibition of IDH1 (R132).
  • Brain-penetrant, enabling central nervous system studies.
  • Supplied as 10 mM solution in DMSO and as solid packs for flexibility.
  • High purity for research-grade applications.
  • Defined storage conditions to preserve stability.
Catalog No. 50-002-42503 Supplier Medchemexpress LLC Supplier No. HY10403610MM 1ML
IDH-305 is an orally available, mutant-selective, brain-penetrant inhibitor of isocitrate dehydrogenase 1 (IDH1) that targets R132 mutations. It is provided for research use in biochemical and cellular assays and is supplied as a 10 mM solution in DMSO (1 mL) or as solid quantities.
  • Potent, mutant-selective inhibition of IDH1 (R132).
  • Brain-penetrant, enabling central nervous system studies.
  • Supplied as 10 mM solution in DMSO and as solid packs for flexibility.
  • High purity for research-grade applications.
  • Defined storage conditions to preserve stability.