Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (4R)-4-(2-Hydroxyethyl)-2,2-Dimethyl-1,3-Dioxolane

Catalog No. 3310745G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-107-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 33-107-45G Supplier Sigma Aldrich Supplier No. 3310745G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(4R)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane can be used:To introduce a chiral center selectively in the carbon chain C10 during the total synthesis of protectin D1.As a starting material in the total synthesis of an anti-diabetic molecule cytopiloyne.As a starting material in the preparation of 4-15N-amino-2-butane-1,2-diol, which is used to synthesize 15N-labeled oligodeoxynucleotides.

Specifications

CAS 17455-25-3
Linear Formula TiOSO4 -+ xH2SO4 -+ yH2O
Refractive Index n20/D 1.4390
MDL Number MFCD00150565
Quantity 5 g
Synonym 2-[(4R)-2,2-Dimethyl-1,3-dioxolan-4-yl]ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.