Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (4R,5R)-2,2-Dimethyl-Alpha,Alpha,Alpha',Alpha'-Tetraphenyldioxolane-4,5-Dimethanol

Catalog No. 2650041G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-500-41G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26-500-41G Supplier Sigma Aldrich Supplier No. 2650041G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(4R,5R)-2,2-Dimethyl-Alpha,Alpha,Alpha',Alpha'-Tetraphenyldioxolane-4,5-Dimethanol

Specifications

CAS 73624-47-2
Density 0.896 g/mL at 25°C
Molecular Formula C18H31B
Linear Formula C18H31B
MDL Number MFCD00074776
Quantity 100 mL
Molecular Weight (g/mol) 258.25 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.