Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4R,5R)-5-(2-chlorophenyl)-4-[5-(2-phenylethynyl)pyridin-3-yl]-1,3-oxazolidin-2-one | 1375752-78-5 | 99.9% | 1 MG
SDP

Catalog No. 5000232165
Encompass_Preferred

BMS-984923 is a small-molecule silent allosteric modulator (SAM) of the metabotropic glutamate receptor 5 (mGluR5), used in research to probe PrPC-mGluR5 interactions and mechanisms related to neurodegeneration. It is supplied as a high-purity solid and as a DMSO solution for in vitro and preclinical studies.

  • High purity: 99.94% (ee 100%).
  • Molecular weight 374.82 g/mol; formula C22H15ClN2O2.
  • Appearance: solid, off-white to light yellow.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available pack sizes include 1 mg solid and 10 mM 1 mL solution in DMSO.

Catalog No. 50-002-32165 Supplier Medchemexpress LLC Supplier No. HY1225591MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-984923 is a small-molecule silent allosteric modulator (SAM) of the metabotropic glutamate receptor 5 (mGluR5), used in research to probe PrPC-mGluR5 interactions and mechanisms related to neurodegeneration. It is supplied as a high-purity solid and as a DMSO solution for in vitro and preclinical studies.

  • High purity: 99.94% (ee 100%).
  • Molecular weight 374.82 g/mol; formula C22H15ClN2O2.
  • Appearance: solid, off-white to light yellow.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available pack sizes include 1 mg solid and 10 mM 1 mL solution in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.