Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4R,7R)-16-chloro-14-fluoro-15-(2-fluoro-6-hydroxyphenyl)-4,9-dimethyl-5-prop-2-enoyl | 2349393-21-9 | 98.5% | 498.91 g/mol | C25H21ClF2N4O3 | 10MM 1ML
SDP

Catalog No. 5000235909
Encompass_Preferred

KRAS G12C inhibitor 15 is a small-molecule covalent inhibitor selective for the KRAS G12C mutant, reported in patent WO2019110751A1 (compound 22). The compound shows potent biochemical activity (IC50 = 5 nM) and is supplied for research use as a high-purity solid or as a ready-to-use DMSO solution.

  • Potent KRAS G12C inhibitor with IC50 = 5 nM.
  • Molecular formula C25H21ClF2N4O3 and molecular weight 498.91 g/mol.
  • Purity 98.5% (HPLC).
  • Available as mg-scale powder (1-100 mg); larger quantities available by quote.
  • Offered as 10 mM solution in DMSO, 1 mL, ready for use.
  • Product data sheet and safety data sheet available for download.

Catalog No. 50-002-35909 Supplier Medchemexpress LLC Supplier No. HY12587310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KRAS G12C inhibitor 15 is a small-molecule covalent inhibitor selective for the KRAS G12C mutant, reported in patent WO2019110751A1 (compound 22). The compound shows potent biochemical activity (IC50 = 5 nM) and is supplied for research use as a high-purity solid or as a ready-to-use DMSO solution.

  • Potent KRAS G12C inhibitor with IC50 = 5 nM.
  • Molecular formula C25H21ClF2N4O3 and molecular weight 498.91 g/mol.
  • Purity 98.5% (HPLC).
  • Available as mg-scale powder (1-100 mg); larger quantities available by quote.
  • Offered as 10 mM solution in DMSO, 1 mL, ready for use.
  • Product data sheet and safety data sheet available for download.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.