Learn More
Medchemexpress LLC (4S)-1-Boc-4-amino-D-proline | 132622-78-7 | MFCD08704543 | ≥97.0% | 230.26 | C10H18N2O4 | 100 MG
SDP
Catalog No. 5000300869
Encompass_Preferred
(4S)-1-Boc-4-amino-D-proline is a Boc-protected derivative of D-proline used as an amino acid building block and intermediate in organic and peptide synthesis. Supplied for research use, it provides a protected primary amine and defined stereochemistry to support selective transformations and peptide assembly.
  • Boc-protected amine for orthogonal deprotection
  • D-configuration (4S) for stereochemical control
  • High purity suitable for research synthesis
  • Molecular weight 230.26 g/mol for predictable stoichiometry
  • Available in small laboratory pack sizes for quick screening
Catalog No. 50-003-00869 Supplier Medchemexpress LLC Supplier No. HYW019205100MG
(4S)-1-Boc-4-amino-D-proline is a Boc-protected derivative of D-proline used as an amino acid building block and intermediate in organic and peptide synthesis. Supplied for research use, it provides a protected primary amine and defined stereochemistry to support selective transformations and peptide assembly.
  • Boc-protected amine for orthogonal deprotection
  • D-configuration (4S) for stereochemical control
  • High purity suitable for research synthesis
  • Molecular weight 230.26 g/mol for predictable stoichiometry
  • Available in small laboratory pack sizes for quick screening