Learn More
Medchemexpress LLC (4S)-1-Boc-4-fluoro-D-proline | 681128-50-7 | 97.0% | 233.24 g·mol⁻¹ | C10H16FNO4 | 25 G
SDP
Catalog No. 5000303034
Encompass_Preferred
(4S)-1-Boc-4-fluoro-D-proline is a Boc-protected, fluoro-substituted D-proline derivative used as a synthetic building block in organic and medicinal chemistry. It supplies a stereodefined fluorinated proline residue for incorporation into peptides and small molecules, enabling conformation control and exploration of structure-activity relationships.
  • Boc-protected amino acid compatible with standard peptide coupling chemistry.
  • Fluorine substitution at the 4-position to influence conformational preference and metabolic stability.
  • D-configuration (4S) provides stereochemical control for peptide design.
  • High chemical purity suitable for synthesis applications.
  • Available in gram-scale quantities for preparative synthesis.
Catalog No. 50-003-03034 Supplier Medchemexpress LLC Supplier No. 25GHYW021482
(4S)-1-Boc-4-fluoro-D-proline is a Boc-protected, fluoro-substituted D-proline derivative used as a synthetic building block in organic and medicinal chemistry. It supplies a stereodefined fluorinated proline residue for incorporation into peptides and small molecules, enabling conformation control and exploration of structure-activity relationships.
  • Boc-protected amino acid compatible with standard peptide coupling chemistry.
  • Fluorine substitution at the 4-position to influence conformational preference and metabolic stability.
  • D-configuration (4S) provides stereochemical control for peptide design.
  • High chemical purity suitable for synthesis applications.
  • Available in gram-scale quantities for preparative synthesis.