Learn More
Medchemexpress LLC (4S)-1-Boc-4-fluoro-D-proline | 681128-50-7 | MFCD08706411 | 97.0% | 233.24 | C10H16FNO4 | 1 G
SDP
Catalog No. 5000300960
Encompass_Preferred
(4S)-1-Boc-4-fluoro-D-proline is a Boc-protected, fluorinated D-proline derivative used as a building block in organic synthesis and peptide chemistry. It serves as an intermediate for preparing peptidomimetics and other fluorinated analogs where stereochemistry and fluorine incorporation are important.
  • Boc-protected fluorinated D-proline for synthetic versatility.
  • High reported purity suitable for research applications.
  • Defined stereochemistry (4S) for stereospecific synthesis.
  • Useful in peptide and peptidomimetic construction.
  • Available in small-scale sizes appropriate for lab use.
Catalog No. 50-003-00960 Supplier Medchemexpress LLC Supplier No. HYW0214821G
(4S)-1-Boc-4-fluoro-D-proline is a Boc-protected, fluorinated D-proline derivative used as a building block in organic synthesis and peptide chemistry. It serves as an intermediate for preparing peptidomimetics and other fluorinated analogs where stereochemistry and fluorine incorporation are important.
  • Boc-protected fluorinated D-proline for synthetic versatility.
  • High reported purity suitable for research applications.
  • Defined stereochemistry (4S) for stereospecific synthesis.
  • Useful in peptide and peptidomimetic construction.
  • Available in small-scale sizes appropriate for lab use.