Learn More
Medchemexpress LLC (4S)-1-Boc-4-fluoro-D-proline | 681128-50-7 | MFCD08706411 | 97.0% | 233.24 g·mol⁻1 | C10H16FNO4 | 10 G
SDP
Catalog No. 5000300491
Encompass_Preferred
(4S)-1-Boc-4-fluoro-D-proline is an N-Boc protected, 4-fluoro substituted D-proline derivative used as a synthetic building block in medicinal chemistry and peptide synthesis. It is employed as an amino acid scaffold for preparing peptidomimetics, modified peptides, and small-molecule intermediates.
  • N-Boc protected 4-fluoro D-proline scaffold for synthesis.
  • Suitable for peptide synthesis and peptidomimetic preparation.
  • High purity for research applications (97.0%).
  • Well-characterized molecular properties: C10H16FNO4, 233.24 g·mol⁻1.
  • Available in multiple pack sizes for synthesis scale flexibility.
Catalog No. 50-003-00491 Supplier Medchemexpress LLC Supplier No. HYW02148210G
(4S)-1-Boc-4-fluoro-D-proline is an N-Boc protected, 4-fluoro substituted D-proline derivative used as a synthetic building block in medicinal chemistry and peptide synthesis. It is employed as an amino acid scaffold for preparing peptidomimetics, modified peptides, and small-molecule intermediates.
  • N-Boc protected 4-fluoro D-proline scaffold for synthesis.
  • Suitable for peptide synthesis and peptidomimetic preparation.
  • High purity for research applications (97.0%).
  • Well-characterized molecular properties: C10H16FNO4, 233.24 g·mol⁻1.
  • Available in multiple pack sizes for synthesis scale flexibility.