Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4S)-1-Boc-4-fluoro-D-proline | 681128-50-7 | MFCD08706411 | 97.0% | 233.24 g/mol | C10H16FNO4 | 5 G
SDP

Catalog No. 5000300488
Encompass_Preferred

(4S)-1-Boc-4-fluoro-D-proline is a Boc-protected, fluorinated D-proline derivative used as a building block in peptide and small-molecule synthesis. Provided as a solid powder, it enables incorporation of a 4-fluoro D-proline residue to modulate conformation and physicochemical properties during medicinal chemistry and peptide design.

  • Boc-protected amino acid enabling selective coupling in synthesis.
  • Contains a 4-fluoro D-proline residue for altered sterics and polarity.
  • High purity (97.0%) suitable for research synthesis.
  • Molecular weight 233.24 g/mol; formula C10H16FNO4.
  • Supplied as a solid, white to off-white, available in gram-scale quantities.
  • Stable under recommended storage conditions for long-term use.

Catalog No. 50-003-00488 Supplier Medchemexpress LLC Supplier No. HYW0214825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(4S)-1-Boc-4-fluoro-D-proline is a Boc-protected, fluorinated D-proline derivative used as a building block in peptide and small-molecule synthesis. Provided as a solid powder, it enables incorporation of a 4-fluoro D-proline residue to modulate conformation and physicochemical properties during medicinal chemistry and peptide design.

  • Boc-protected amino acid enabling selective coupling in synthesis.
  • Contains a 4-fluoro D-proline residue for altered sterics and polarity.
  • High purity (97.0%) suitable for research synthesis.
  • Molecular weight 233.24 g/mol; formula C10H16FNO4.
  • Supplied as a solid, white to off-white, available in gram-scale quantities.
  • Stable under recommended storage conditions for long-term use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.