Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4S)-2-amino-N-ethyl-4,5-dihydro-N-phenyl-4-oxazolemethanamine | 1048346-74-2 | 99.4% | 219.29 g/mol | C12H17N3O | 5 MG
SDP

Catalog No. 5000264455
Encompass_Preferred

RO5166017 is a potent, orally active agonist of the trace amine-associated receptor 1 (TAAR1) used as a research tool in preclinical pharmacology. It exhibits low-nanomolar activity in rodent TAAR1 and measurable activity at human TAAR1, and the product datasheet provides purity, solubility, and storage guidance for experimental use.

  • Orally active TAAR1 agonist with nanomolar potency.
  • Binding Ki values: mouse 1.9 nM, rat 2.7 nM, human 31 nM, cynomolgus 24 nM.
  • High purity, reported as about 99.4%.
  • Molecular formula C12H17N3O; molecular weight 219.29 g/mol.
  • Soluble in DMSO (25 mg/mL); in vivo formulation options and concentrations provided.
  • Storage recommendations: -20°C for solid; in solvent, -80°C for long-term storage.

Catalog No. 50-002-64455 Supplier Medchemexpress LLC Supplier No. HY126995MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RO5166017 is a potent, orally active agonist of the trace amine-associated receptor 1 (TAAR1) used as a research tool in preclinical pharmacology. It exhibits low-nanomolar activity in rodent TAAR1 and measurable activity at human TAAR1, and the product datasheet provides purity, solubility, and storage guidance for experimental use.

  • Orally active TAAR1 agonist with nanomolar potency.
  • Binding Ki values: mouse 1.9 nM, rat 2.7 nM, human 31 nM, cynomolgus 24 nM.
  • High purity, reported as about 99.4%.
  • Molecular formula C12H17N3O; molecular weight 219.29 g/mol.
  • Soluble in DMSO (25 mg/mL); in vivo formulation options and concentrations provided.
  • Storage recommendations: -20°C for solid; in solvent, -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.