Learn More
Medchemexpress LLC (4S)-4-(2-methylpropyl)pyrrolidin-2-one | 181289-23-6 | MFCD19217624 | 100.0% | 141.21 | C8H15NO | 250 MG
SDP
Catalog No. 5000303005
Encompass_Preferred
Pregabalin lactam is the S-isomer lactam impurity of pregabalin supplied as an analytical reference material for research, quality control, and impurity profiling. It is typically provided as an off-white to light yellow solid-liquid mixture with specification information for analytical use.
  • High purity (99.99%).
  • Molecular weight 141.21 g/mol and formula C8H15NO for precise identification.
  • Suitable for analytical reference, method development, and impurity profiling.
  • Available in small package sizes suitable for laboratory use.
Catalog No. 50-003-03005 Supplier Medchemexpress LLC Supplier No. 250MGHYW038877
Pregabalin lactam is the S-isomer lactam impurity of pregabalin supplied as an analytical reference material for research, quality control, and impurity profiling. It is typically provided as an off-white to light yellow solid-liquid mixture with specification information for analytical use.
  • High purity (99.99%).
  • Molecular weight 141.21 g/mol and formula C8H15NO for precise identification.
  • Suitable for analytical reference, method development, and impurity profiling.
  • Available in small package sizes suitable for laboratory use.