Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4S)-4-(3-bromophenyl)-4,5-dihydro-2-(methylthio)thiazole-5-carboxylic acid | 330203-01-5 | 99.0% | 422.32 g·mol⁻¹ | C17H12BrNO3S2 | 1 ML
SDP

Catalog No. 5000245186
Encompass_Preferred

TC HSD 21 is a potent, selective inhibitor of 17β-hydroxysteroid dehydrogenase type 3 (17β-HSD3) with an IC50 of 14 nM. It is supplied as a 10 mM solution in DMSO (1 mL vial) and has a molecular formula of C17H12BrNO3S2, molecular weight 422.32 g·mol⁻¹, and purity 99.0%.

  • Inhibits 17β-HSD3 with an IC50 of 14 nM.
  • Shows excellent selectivity over other 17β-HSD isoenzymes and nuclear receptors.
  • Supplied as a 10 mM solution in DMSO (1 mL) for ready-to-use assays.
  • High purity (99.0%) for reliable biochemical studies.
  • Molecular weight 422.32 g·mol⁻¹ and molecular formula C17H12BrNO3S2.
  • Recommended storage: powder -20°C for long term; in solvent -80°C for up to 6 months.

Catalog No. 50-002-45186 Supplier Medchemexpress LLC Supplier No. HY10339410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TC HSD 21 is a potent, selective inhibitor of 17β-hydroxysteroid dehydrogenase type 3 (17β-HSD3) with an IC50 of 14 nM. It is supplied as a 10 mM solution in DMSO (1 mL vial) and has a molecular formula of C17H12BrNO3S2, molecular weight 422.32 g·mol⁻¹, and purity 99.0%.

  • Inhibits 17β-HSD3 with an IC50 of 14 nM.
  • Shows excellent selectivity over other 17β-HSD isoenzymes and nuclear receptors.
  • Supplied as a 10 mM solution in DMSO (1 mL) for ready-to-use assays.
  • High purity (99.0%) for reliable biochemical studies.
  • Molecular weight 422.32 g·mol⁻¹ and molecular formula C17H12BrNO3S2.
  • Recommended storage: powder -20°C for long term; in solvent -80°C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.