Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (4S,4'S)-(-)-2,2'-(3-Pentylidene)Bis(4-Isopropyloxazoline)

Catalog No. 650897500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-089-7500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 65-089-7500MG Supplier Sigma Aldrich Supplier No. 650897500MG
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(4S,4'saS)-(-)-2,2'sa-(3-Pentylidene)bis(4-isopropyloxazoline) is a bisoxazoline that can be used as a ligand:To prepare [11C]carboxylic acids, [11C]esters and [11C]amides using boronic acid esters and 11CO2.In one of the key synthetic steps for the total synthesis of tatanan A and 3-epi-tatanan A.To prepare copper(I) complex and used in a mass spectrometric method to study its chiral selectivity with different substrates.

Specifications

CAS 10090-05-8
Density 1.09 g/mL at 25°C
Boiling Point 103°C to 104°C (25 mmHg, lit.)
Molecular Formula C4H12O3SSi
Linear Formula H3CSO3OSi(CH3)3
Refractive Index n20/D 1.4710
MDL Number MFCD00042815
Quantity 500 mg
Synonym (4S,4'S)-(-)-2,2'-(1-Ethylpropylidene)bis(4,5-dihydro-4-isopropyloxazole); [S-(R,R)]-(-)-2,2'-(1-Ethylpropylidene)bis(4,5-dihydro-4-isopropyloxazole)
Molecular Weight (g/mol) 168.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.