Learn More
Medchemexpress LLC (4S)-GLP-1 receptor agonist 14 | 2758659-09-3 | 99.6% | 880.94 | 5 MG
SDP
Catalog No. 5000391431
Encompass_Preferred
(4S)-GLP-1 receptor agonist 14 is a potent and orally active GLP-1 receptor agonist with an EC50 ≤ 20 nM. It is intended for research applications related to diabetes, obesity, metabolic diseases, cardiovascular diseases, liver diseases, and nonalcoholic steatohepatitis (NASH), as well as other conditions associated with the GLP-1 receptor. In vitro, it inhibits OATP1B1 with an IC50 < 1 μM in HEK293 cells. In vivo, a single oral dose at 5 mg/kg in rats demonstrates good pharmacokinetic parameters, with a Cmax of 1700 ng/mL, AUC of 10193 ng·h/mL, and an oral bioavailability (F) of 90%.
  • Potent and orally active GLP-1 receptor agonist
  • EC50 ≤ 20 nM
  • Inhibits OATP1B1 with an IC50 < 1 μM in HEK293 cells
  • Exhibits good pharmacokinetic parameters in rats
  • Suitable for research on diabetes, obesity, metabolic diseases, cardiovascular diseases, liver diseases, and nonalcoholic steatohepatitis (NASH)
Catalog No. 50-003-91431 Supplier Medchemexpress LLC Supplier No. HY1647745MG
Add to Cart
Add to Cart
(4S)-GLP-1 receptor agonist 14 is a potent and orally active GLP-1 receptor agonist with an EC50 ≤ 20 nM. It is intended for research applications related to diabetes, obesity, metabolic diseases, cardiovascular diseases, liver diseases, and nonalcoholic steatohepatitis (NASH), as well as other conditions associated with the GLP-1 receptor. In vitro, it inhibits OATP1B1 with an IC50 < 1 μM in HEK293 cells. In vivo, a single oral dose at 5 mg/kg in rats demonstrates good pharmacokinetic parameters, with a Cmax of 1700 ng/mL, AUC of 10193 ng·h/mL, and an oral bioavailability (F) of 90%.
  • Potent and orally active GLP-1 receptor agonist
  • EC50 ≤ 20 nM
  • Inhibits OATP1B1 with an IC50 < 1 μM in HEK293 cells
  • Exhibits good pharmacokinetic parameters in rats
  • Suitable for research on diabetes, obesity, metabolic diseases, cardiovascular diseases, liver diseases, and nonalcoholic steatohepatitis (NASH)