Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4S)-GLP-1 receptor agonist 14 | 2758659-09-3 | 99.6% | 880.94 | C48H46F2N10O5 | 10 MG
SDP

Catalog No. 5000289321
Encompass_Preferred

(4S)-GLP-1 receptor agonist 14 is a research-grade small-molecule modulator of the glucagon-like peptide-1 (GLP-1) receptor supplied as a solid for in vitro and preclinical research use. Not for human or therapeutic use.

  • High purity (99.6%).
  • Molecular formula C48H46F2N10O5.
  • Molecular weight 880.94 g/mol.
  • Appearance solid.
  • Solubility in DMSO 125 mg/mL (141.89 mM); ultrasonic recommended.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solution -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-89321 Supplier Medchemexpress LLC Supplier No. HY16477410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(4S)-GLP-1 receptor agonist 14 is a research-grade small-molecule modulator of the glucagon-like peptide-1 (GLP-1) receptor supplied as a solid for in vitro and preclinical research use. Not for human or therapeutic use.

  • High purity (99.6%).
  • Molecular formula C48H46F2N10O5.
  • Molecular weight 880.94 g/mol.
  • Appearance solid.
  • Solubility in DMSO 125 mg/mL (141.89 mM); ultrasonic recommended.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solution -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.