Learn More
Medchemexpress LLC (4S)-PROTAC SOS1 degrader-1 | 2913176-81-3 | C57H76ClFN10O4S | 50 MG
SDP
Catalog No. 5000354818
Encompass_Preferred
(4S)-PROTAC SOS1 degrader-1 is a potent PROTAC SOS1 degrader. It decreases the expression of pERK and RAS-GTP levels in a dose-dependent manner. It also significantly inhibits tumor growth in vivo.
  • Shows potent SOS1 degradation activity with 56.2% and 92.5% SOS1 protein degradation at 0.1 and 1 μM, respectively, in NCI-H358 cells.
  • Degrades SOS1 in a dose- and time-dependent manner in NCI-H358 cells (24 h incubation).
  • Does not induce SOS2 and KRAS degradation in NCI-H358 cells (9.8-2500 nM; 24 h).
  • Decreases the expression of pERK and RAS-GTP levels in NCI-H358 cells in a dose-dependent manner (7.8-2000 nM; 24 h).
  • Significantly inhibits tumor growth in vivo.
Catalog No. 50-003-54818 Supplier Medchemexpress LLC Supplier No. HY144657A50MG
Add to Cart
Add to Cart
(4S)-PROTAC SOS1 degrader-1 is a potent PROTAC SOS1 degrader. It decreases the expression of pERK and RAS-GTP levels in a dose-dependent manner. It also significantly inhibits tumor growth in vivo.
  • Shows potent SOS1 degradation activity with 56.2% and 92.5% SOS1 protein degradation at 0.1 and 1 μM, respectively, in NCI-H358 cells.
  • Degrades SOS1 in a dose- and time-dependent manner in NCI-H358 cells (24 h incubation).
  • Does not induce SOS2 and KRAS degradation in NCI-H358 cells (9.8-2500 nM; 24 h).
  • Decreases the expression of pERK and RAS-GTP levels in NCI-H358 cells in a dose-dependent manner (7.8-2000 nM; 24 h).
  • Significantly inhibits tumor growth in vivo.