Learn More
Medchemexpress LLC (4S)-protac SOS1 degrader-1 diTFA | 00-00-0 | 98.1% | C57H76ClFN10O4S | 10MG
SDP
Catalog No. 5000258406
Encompass_Preferred
(4S)-PROTAC SOS1 degrader-1 (diTFA) is a PROTAC small molecule supplied as the di-trifluoroacetate salt for research use. It induces targeted degradation of Son of Sevenless 1 (SOS1), reduces pERK and RAS-GTP levels in a dose-dependent manner, and has been reported to inhibit tumor growth in vivo. Product documentation, including COA and SDS, is available.
Catalog No. 50-002-58406 Supplier Medchemexpress LLC Supplier No. HY144657A10MG
(4S)-PROTAC SOS1 degrader-1 (diTFA) is a PROTAC small molecule supplied as the di-trifluoroacetate salt for research use. It induces targeted degradation of Son of Sevenless 1 (SOS1), reduces pERK and RAS-GTP levels in a dose-dependent manner, and has been reported to inhibit tumor growth in vivo. Product documentation, including COA and SDS, is available.