Learn More
Apexbio Technology LLC 4SC-202, 5mg. Cas: 1186222-89-8 MFCD: MFCD25976779. Class I HDAC inhibitor
SDP
Supplier:  Apexbio Technology LLC B41065
Encompass_Preferred
4SC-202 is a selective, potent and orally available inhibitor of histone deacetylases (HDAC) specific for class I HDAC isoenzymes with an IC50 value of about 1M [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5353