Learn More
TARGETMOL CHEMICALS INC 4SC-203 5MG
SDP
Catalog No. 502576201
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. 4SC-203 is a multikinase inhibitor with potential antineoplastic activity. 4SC-203 selectively inhibits FMS-related tyrosine kinase 3 (FLT3/STK1) FLT3 mutated forms and vascular endothelial growth factor receptors (VEGFRs). purity: 99%
Catalog No. 50-257-6201 Supplier TARGETMOL CHEMICALS INC Supplier No. T94735MG
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. 4SC-203 is a multikinase inhibitor with potential antineoplastic activity. 4SC-203 selectively inhibits FMS-related tyrosine kinase 3 (FLT3/STK1) FLT3 mutated forms and vascular endothelial growth factor receptors (VEGFRs). purity: 99%