Learn More
Medchemexpress LLC 4SC-203 | 895533-09-2 | 98.98% | 642.77 | 5 MG
SDP
Catalog No. 5000400502
Encompass_Preferred
4SC-203 is a potent multikinase inhibitor with potential antineoplastic activity. It selectively targets FLT3/STK1, FLT3 mutated forms, and VEGFRs.
  • Potent multikinase inhibitor.
  • Potential antineoplastic activity.
  • Selectively inhibits FLT3/STK1, FLT3 mutated forms, and VEGFRs.
Catalog No. 50-004-00502 Supplier Medchemexpress LLC Supplier No. HY198975MG
4SC-203 is a potent multikinase inhibitor with potential antineoplastic activity. It selectively targets FLT3/STK1, FLT3 mutated forms, and VEGFRs.
  • Potent multikinase inhibitor.
  • Potential antineoplastic activity.
  • Selectively inhibits FLT3/STK1, FLT3 mutated forms, and VEGFRs.