Learn More
Medchemexpress LLC 4SC-203 | 895533-09-2 | 99.0% | 642.77 | 50 MG
SDP
Catalog No. 5000401326
Encompass_Preferred
4SC-203 is a potent multikinase inhibitor that has potential antineoplastic activity. It selectively inhibits FLT3/STK1, mutated forms of FLT3, and VEGFRs.
  • Potent multikinase inhibitor.
  • Exhibits potential antineoplastic activity.
  • Selectively targets FLT3/STK1.
  • Selectively targets FLT3 mutated forms.
  • Selectively targets VEGFRs.
Catalog No. 50-004-01326 Supplier Medchemexpress LLC Supplier No. HY1989750MG
4SC-203 is a potent multikinase inhibitor that has potential antineoplastic activity. It selectively inhibits FLT3/STK1, mutated forms of FLT3, and VEGFRs.
  • Potent multikinase inhibitor.
  • Exhibits potential antineoplastic activity.
  • Selectively targets FLT3/STK1.
  • Selectively targets FLT3 mutated forms.
  • Selectively targets VEGFRs.