Learn More
ATCC 4T1; Mammary Tumor; Mouse (Mus musculus)
Catalog No. 502381807
Encompass
The tumor growth and metastatic spread of 4T1 cells in BALB/c mice very closely mimic human breast cancer. This tumor-derived cell line is an animal model for stage IV human breast cancer. 4T1-induced tumors can be used as a post-operative model as well as a nonsurgical model because the 4T1-induced tumor metastasizes spontaneously in both models with similar kinetics.
Catalog No. 50-238-1807 Supplier ATCC Supplier No. CRL2539
The tumor growth and metastatic spread of 4T1 cells in BALB/c mice very closely mimic human breast cancer. This tumor-derived cell line is an animal model for stage IV human breast cancer. 4T1-induced tumors can be used as a post-operative model as well as a nonsurgical model because the 4T1-induced tumor metastasizes spontaneously in both models with similar kinetics.