Learn More
ATCC 4T1.13; Breast Carcinoma; Mouse (Mus musculus)
Catalog No. 502384044
Encompass
4T1.3 is a cell line with epithelial-like morphology that was isolated in 2004 from the breast of a Balb/cfC3H mouse, mimicking Stage IV breast cancer in patients. Identified as a single cell clone of 4T1 (ATCC CRL-2539) with high propensity to metastasize spontaneously to bone, causing fractures and/or hind limb paralysis. Primary tumor growth properties similar to those of 4T1. It has applications for cancer research and is used for the analysis of genes that regulate metastasis of breast cancer and/or drugs that might reduce spontaneous metastasis in immune-competent Balb/c mice.
Catalog No. 50-238-4044 Supplier ATCC Supplier No. CRL3407
4T1.3 is a cell line with epithelial-like morphology that was isolated in 2004 from the breast of a Balb/cfC3H mouse, mimicking Stage IV breast cancer in patients. Identified as a single cell clone of 4T1 (ATCC CRL-2539) with high propensity to metastasize spontaneously to bone, causing fractures and/or hind limb paralysis. Primary tumor growth properties similar to those of 4T1. It has applications for cancer research and is used for the analysis of genes that regulate metastasis of breast cancer and/or drugs that might reduce spontaneous metastasis in immune-competent Balb/c mice.