Learn More
Description
- Features detailed bronchial bifurcations and over 20 visible anatomical landmarks for accurate anatomy training.
- Optional Add-on designed to enhance the functionality of the P73 and P73D airway management trainers.
- Features detailed bronchial bifurcations and over 20 visible anatomical landmarks for accurate anatomy training.
- Provides realistic practice for medical professionals and students to develop essential airway management skills.
- Includes tracheal membrane, cartilage ring notches, aortic arch crossover curvature, carina, and left main bronchus pathway.
- The bronchial structure features 28 branching points, clearly visualizing the third and fourth level bronchi.
- Main Bronchi includes left and right main bronchi. The right bronchus branches into upper, middle, and lower lobes, while the left bronchus branches into upper and lower lobes.
- The model provides a detailed view of the segmental bronchi and their tertiary branches.
- Supports flexible bronchoscope use for airway examination,lavage, and foreign body removal.
- Simulate partial or complete airway obstruction through manual compression or clamping for advanced training scenarios, clamps should not be used for Long-term storage.
- Allows the placement of solid, liquid, or mixed foreign bodies in the airway for realistic training scenarios.
- Supports the extraction of objects up to three times the tracheal diameter, providing challenging practice for advanced foreign body retrieval techniques.
- Made of biodegradable latex, the module is easy to maintain and can be flushed with water for thorough internal cleaning.
- Serves as an essential tool for developing clinical skills in a safe and controlled environment.
- Avoid prolonged exposure to liquids, foreign objects, chemicals, or high temperatures, as these may damage the module.
- Clean thoroughly after each use and keep the module dry to maintain durability.
- Avoid exposure to UV light, as it may damage the material.
Specifications
Specifications
| Product Type | Manikin |
| For Use With (Application) | Medical Training |
| Color | Red |
| Gender | Male, Female |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


