Learn More
Thomas Scientific PCR Seal; clear adhesive film, strong adhesive, 100 sheets (135 x 80 mm)
SDP
Catalog No. NC1172964
Encompass
Clear adhesive film, strong adhesive, peelable, suitable for PCR and optical applications Our PCR Seal is a durable transparent polyester film with a strong adhesive layer. The seal enables a high integrity and efficiently prevents sample evaporation. It is recommended for PCR, qPCR, and other optical applications (e.g. fluorescence or colorimetric measurements) as well as sample storage. The PCR Seal is also available in two flexible formats with perforated sheets, to enable tearing into 8 well and 12 well strips, respectively as well as in half plate size format to be used with our FrameStar® 192 Well Semi-Skirted PCR Plate. This allows for sealing of complete 96 well plates, but also individual or multiple Tear-A-Way™ or Break-A-Way™ strips, perfectly complementing these products.
Catalog No. NC1172964 Supplier Thomas Scientific Supplier No. 1149J07
Clear adhesive film, strong adhesive, peelable, suitable for PCR and optical applications Our PCR Seal is a durable transparent polyester film with a strong adhesive layer. The seal enables a high integrity and efficiently prevents sample evaporation. It is recommended for PCR, qPCR, and other optical applications (e.g. fluorescence or colorimetric measurements) as well as sample storage. The PCR Seal is also available in two flexible formats with perforated sheets, to enable tearing into 8 well and 12 well strips, respectively as well as in half plate size format to be used with our FrameStar® 192 Well Semi-Skirted PCR Plate. This allows for sealing of complete 96 well plates, but also individual or multiple Tear-A-Way™ or Break-A-Way™ strips, perfectly complementing these products.