Learn More
Cayman Chemical 4-tert-Octylphenol diethoxylate, Purity: ≥98%, CAS: 2315-61-9, 10 mg
SDP
Catalog No. 502965656
Encompass_Preferred
A degradation product of the surfactant, Triton X that produces weak estrogenic effects in animal and human research studies
Catalog No. 50-296-5656 Supplier Cayman Chemical Supplier No. 1865510 MG
A degradation product of the surfactant, Triton X that produces weak estrogenic effects in animal and human research studies