Learn More
Cayman Chemical 4-tert-Octylphenol monoethoxylate, Purity: ≥98%, CAS: 2315-67-5, 10 mg
SDP
Catalog No. 502919293
Encompass_Preferred
An alkylphenolethoxylate and a degradation product of non-ionic surfactants; a non-steroidal estrogen; found in wastewater effluent
Catalog No. 50-291-9293 Supplier Cayman Chemical Supplier No. 3736610 MG
An alkylphenolethoxylate and a degradation product of non-ionic surfactants; a non-steroidal estrogen; found in wastewater effluent