Learn More
Apexbio Technology LLC 4μ8C 14003-96-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061739
Encompass_Preferred
4 8C (CAS 14003-96-4) is a small molecule inhibitor targeting the RNase activity of inositol-requiring enzyme 1 alpha (IRE1 ) an important transmembrane serine/threonine kinase involved in the unfolded protein response (UPR) By selectively blocking IRE1-mediated RNase function 4 8C inhibits downstream UPR gene activation in cell lines exposed to hypoxia or endoplasmic reticulum (ER) stressors including HCT116 colon carcinoma and KP4 pancreatic cancer cells without affecting proliferation or clonogenic survival under these conditions Due to its selective action 4 8C serves as a valuable tool in research investigating ER stress pathways and their roles in cellular adaptation and disease pathology
Catalog No. 50-000-61739 Supplier Apexbio Technology LLC Supplier No. B18745.1
4 8C (CAS 14003-96-4) is a small molecule inhibitor targeting the RNase activity of inositol-requiring enzyme 1 alpha (IRE1 ) an important transmembrane serine/threonine kinase involved in the unfolded protein response (UPR) By selectively blocking IRE1-mediated RNase function 4 8C inhibits downstream UPR gene activation in cell lines exposed to hypoxia or endoplasmic reticulum (ER) stressors including HCT116 colon carcinoma and KP4 pancreatic cancer cells without affecting proliferation or clonogenic survival under these conditions Due to its selective action 4 8C serves as a valuable tool in research investigating ER stress pathways and their roles in cellular adaptation and disease pathology