Learn More
Electron Microscopy Sciences 4-Way Microtube Rack Assorted
SDP
Catalog No. 501902299
Encompass_Preferred
Create as large a rack as you need by linking together multiple racks. Heavyduty polypropylene racks feature a unique system of tabs and slots, that facilitate easy connection and sturdy fit. Autoclavable. Assorted colors or natural.
Catalog No. 50-190-2299 Supplier Electron Microscopy Sciences Supplier No. 6161500
Create as large a rack as you need by linking together multiple racks. Heavyduty polypropylene racks feature a unique system of tabs and slots, that facilitate easy connection and sturdy fit. Autoclavable. Assorted colors or natural.