Learn More
World Precision Instrument 4-Way Stopcock, Yellow, 2 Female Luer Locks, Male Luer with Spin Lock
SDP
Catalog No. 50822037
Encompass
  • Polycarbonate (medical grade) body
  • Polyethylene handle 
  • 130 PSI
  • 4-way stopcock, luer lock, package of 10
Catalog No. 50-822-037 Supplier World Precision Instrument Supplier No. 1405710
  • Polycarbonate (medical grade) body
  • Polyethylene handle 
  • 130 PSI
  • 4-way stopcock, luer lock, package of 10