Learn More
Electron Microscopy Sciences PEAK 4x Achromatic Magnifier
SDP
Supplier:  Electron Microscopy Sciences 6812804
Encompass_Preferred
Uses high-resolution achromatic lenses whose chromatic aberration has been corrected. With its wide square Field-of-View (45x45 mm) and its interchangeable semi-transparent/ black skirt, this unique magnifier offers you a variety of uses for precision work. Fine focus adjustment with multi-groove screw helicoid system gives you very smooth operation. Neck strap is included.
Catalog No. 50-277-30