Learn More
Genscript Corporation 4X LDS Sample Buffer
Catalog No. NC1395877
Encompass
4X LDS Sample Buffer is used to prepare protein samples for denaturing polyacrylamide gel electrophoresis (PAGE) with SurePAGE™, ExpressPlus™ and most other types of Bis-Tris gels. LDS sample buffer contains lithium dodecyl sulfate with pH at 8.4, which helps reducing the disulfide bonds and ensure optimal protein separation.
Catalog No. NC1395877 Supplier Genscript Corporation Supplier No. M0067610
4X LDS Sample Buffer is used to prepare protein samples for denaturing polyacrylamide gel electrophoresis (PAGE) with SurePAGE™, ExpressPlus™ and most other types of Bis-Tris gels. LDS sample buffer contains lithium dodecyl sulfate with pH at 8.4, which helps reducing the disulfide bonds and ensure optimal protein separation.