Learn More
Vmrd Inc 4X FA Powdered Rinse Buffer
SDP
Catalog No. NC1113472
Encompass
FA Rinse Buffer, pH 9.0. Powder to make one liter of 4x rinse buffer. Used to rinse fluorescent antibody antibody substrate slides.
Catalog No. NC1113472 Supplier Vmrd Inc Supplier No. FARB4X
FA Rinse Buffer, pH 9.0. Powder to make one liter of 4x rinse buffer. Used to rinse fluorescent antibody antibody substrate slides.