Learn More
LICORbio 4X Protein Sample Loading Buffer and PVDF Membrane Kit
Catalog No. NC1943776
Encompass_Preferred
4X Protein Sample Loading Buffer is optimized for use as a loading buffer for protein gel electrophoresis. This orange-colored loading buffer has no dyes that may cause background as compare to other loading buffers with a blue dye component that can increase background. The Immobilon-FL PVDF membrane included in these kits has been tested to ensure a low-background for near-infrared fluorescent Western blot success. This kit contains 0.45 µm; 26.5 cm x 3.75 m roll of Immobilon-FL PVDF membrane and 15 mL 4X Protein Sample Loading Buffer
Catalog No. NC1943776 Supplier LICORbio Supplier No. 92631097
4X Protein Sample Loading Buffer is optimized for use as a loading buffer for protein gel electrophoresis. This orange-colored loading buffer has no dyes that may cause background as compare to other loading buffers with a blue dye component that can increase background. The Immobilon-FL PVDF membrane included in these kits has been tested to ensure a low-background for near-infrared fluorescent Western blot success. This kit contains 0.45 µm; 26.5 cm x 3.75 m roll of Immobilon-FL PVDF membrane and 15 mL 4X Protein Sample Loading Buffer