Learn More
LICORbio 4X Protein Sample Loading Buffer for Western Blots (15 mL)
Catalog No. NC9779096
Encompass_Preferred
4X Protein Sample Loading Buffer is optimized for use as a loading buffer for protein gel electrophoresis. This orange loading buffer is recommended for use with Odyssey Imaging Systems as it does not fluoresce in the 700 nm channel the way blue loading buffers do. The buffer is optimized for use with SDS-PAGE and Tris-Glycine-SDS running buffer.
Catalog No. NC9779096 Supplier LICORbio Supplier No. 92840004
4X Protein Sample Loading Buffer is optimized for use as a loading buffer for protein gel electrophoresis. This orange loading buffer is recommended for use with Odyssey Imaging Systems as it does not fluoresce in the 700 nm channel the way blue loading buffers do. The buffer is optimized for use with SDS-PAGE and Tris-Glycine-SDS running buffer.